Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
This antibody can recognize the heavy chain.
Immunogen sequence: FGYSVALHR QTERQQRYLL LAGAPRELAV PDGYTNRTGA VYLCPLTAHK DDCERMNITV KNDPGHHIIE DMWLGVTVAS QGPAGRVLVC AHRYTQVLWS GSEDQRRMVG KCYVRGNDLE LDSSDDWQTY HNEMCNSNTD YLETGMCQLG TSGGFTQNTV YFGAPGAYNW KGNSYMIQRK EWDLSEYSYK DPEDQGNLYI GYTMQVGSFI LHPKNITIVT GAPRHRHMGA VFLLSQEAGG DLRRRQVLEG SQVGAYFGSA IALADLNNDG WQDLLVGAPY YFERKEEVGG AIYVFMNQAG TSFPAHPSLL LHGPSGSAFG LSVASI (52-376 aa encoded by BC150190)
ITGA3 (INTEGRIN, ALPHA-3), also called CD49C, VLA3 or GAPB3, is a protein that in humans is encoded by the ITGA3 gene. ITGA3 is an integrin alpha subunit which is also a member of the family of cell surface adhesion molecules. ITGA3 is mapped to chromosome 17 and its exact cytogenetic location is 17q21.33. ITGA3 makes up half of the alpha3beta1 integrin duplex that plays a role in neural migration and corticogenesis together with beta-1 subunit. A functional link between DAB1 phosphorylation and ITGA3 signaling drives the timely detachment of migrating neurons from radial glial fibers. Expression of human ITGA3 increased the infectivity of virus for Chinese hamster ovary cells. ITGA3 also contains 13 potential N-glycosylation sites, 2 potential cleavage sites, and the 7 N-terminal repeating units characteristic of ITGAs. Recombinant ITGA3 is expressed as a 150-kD protein as the same size as the native protein by the western blot analysis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha 3 subunit of VLA-3 receptor; Alpha3 integrin; antigen CD49C; antigen identified by monoclonal antibody J143; CD49 antigen-like family member C; CD49c; CD49c antigen; FLJ34631; FLJ34704; FRP-2; Galactoprotein B3; GAPB3; integrin alpha 3; Integrin alpha-3; Integrin alpha3; integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor); testicular tissue protein Li 96; testicular tissue protein Li 98; very late activation protein 3 receptor, alpha-3 subunit; VLA-3 subunit alpha
Gene Aliases: AA407068; CD49C; FRP-2; GAP-B3; GAPB3; ILNEB; ITGA3; MSK18; VCA-2; VL3A; VLA3a
UniProt ID: (Human) P26006, (Mouse) Q62470
Entrez Gene ID: (Human) 3675, (Pig) 100517053, (Mouse) 16400
Molecular Function: integrin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support