Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of total IFNA8.
Interferon alpha-8 Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IFN-alpha-8; interferon alpha type 201; Interferon alpha-8; Interferon alpha-B; interferon alpha-B'; Interferon alpha-B2; interferon, alpha 8; LeIF B; RP11-354P17.14-001
Gene Aliases: IFN-alphaB; IFNA8
UniProt ID: (Human) P32881
Entrez Gene ID: (Human) 3445
Molecular Function: interferon superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support