Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
PA1-26880 detects calf intestinal Alkaline Phosphatase from bovine samples.
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The intestinal alkaline phosphatase gene encodes a digestive brush-border enzyme. This enzyme is upregulated during small intestinal epithelial cell differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Akp2; AP; EC 3.1.3.1; I Kasahara isozyme; intestinal alkaline phosphatase V; PLAP-1; Regan isozyme
Gene Aliases: IAP
Entrez Gene ID: (Bovine) 280993
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support