Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 7
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji.
Predicted reactivity: Mouse (87%), Rat (87%), Bovine (85%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The intestinal alkaline phosphatase gene encodes a digestive brush-border enzyme. This enzyme is upregulated during small intestinal epithelial cell differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Akp2; alkaline phosphatase 6; alkaline phosphomonoesterase; EC 3.1.3.1; glycerophosphatase; I Kasahara isozyme; IAP; intestinal alkaline phosphatase; Intestinal-type alkaline phosphatase; Kasahara isozyme; Regan isozyme
Gene Aliases: 2010001C14Rik; Akp6; ALPI; gIAP; IAP
UniProt ID: (Human) P09923
Entrez Gene ID: (Human) 248, (Mouse) 76768
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support