Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is rat brain tissue lysate.
PA5-34495 can be used with blocking peptide PEP-1537.
JAKMIP1, also known as Marlin-1, is a highly-conserved vertebrate-specific microtubule-associated protein that plays a role in JAK1 signaling and regulate microtubule cytoskeleton rearrangements. JAKMIP1 is expressed predominantly in neuronal and lymphoid cells and colocalizes with microtubules. Multiple isoforms of JAKMIP1 are known to exist: isoforms 1 and 5 are also expressed in liver, lung and skeletal muscle, while isoforms 2-4 are specifically expressed in brain and retina. JAKMIP1 participates in polarized secretion in lymphocytes and it has been suggested to be a novel effector memory gene restraining T cell-mediated cytotoxicity. Dysregulation of JAKMIP1 gene expression is associated with genetic conditions linked to autism in humans.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ31564; GABA-B receptor-binding protein; gamma-aminobutyric acid (GABA-B) receptor binding protein; Jak and microtubule interacting protein 1; Janus kinase and microtubule-interacting protein 1; Marlin-1; Multiple alpha helices and RNA-linker protein 1; Multiple alpha-helices and RNA-linker protein 1; multiple coiled-coil GABABR1-binding protein
Gene Aliases: 5830437M04Rik; C330021K24Rik; GABABRBP; JAKMIP1; JAMIP1; Marlin-1; MARLIN1
UniProt ID: (Human) Q96N16, (Rat) Q3SWS9, (Mouse) Q8BVL9
Entrez Gene ID: (Human) 152789, (Rat) 305434, (Mouse) 76071
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support