Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat and Orangutan
TAOK3 (JIK) is a serine/threonine kinase that is a member of the GCK-like subfamily of the STE20 kinase family. JIK is ubiquitously expressed in human tissues, inhibits JNK and has been implicated in cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTCL-associated antigen HD-CL-09; Cutaneous T-cell lymphoma-associated antigen HD-CL-09; Dendritic cell-derived protein kinase; DKFZp666H245; EC 2.7.11.1; FLJ31808; hKFC-A; JNK/SAPK-inhibitory kinase; Jun kinase-inhibitory kinase; Kinase from chicken homolog A; Serine/threonine-protein kinase TAO3; STE2-like kinase; STE20-like kinase; tao; Thousand and one amino acid protein 3
Gene Aliases: 2900006A08Rik; A130052D22; A430105I05Rik; DPK; JIK; KDS; MAP3K18; TAOK3
UniProt ID: (Human) Q9H2K8, (Mouse) Q8BYC6
Entrez Gene ID: (Human) 51347, (Mouse) 330177
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support