Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The protein encoded by this gene is closely related to MAPK8IP1/IB1/JIP-1, a scaffold protein that is involved in the c-Jun amino-terminal kinase signaling pathway. This protein is expressed in brain and pancreatic cells. It has been shown to interact with, and regulate the activity of MAPK8/JNK1, and MAP2K7/MKK7 kinases. This protein thus is thought to function as a regulator of signal transduction by protein kinase cascade in brain and pancreatic beta-cells. Alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-Jun-amino-terminal kinase-interacting protein 2; homologous to mouse JIP-1; IB 2; IB-2; Islet brain 2; islet-brain 2; Islet-brain-2; JIP 2; JIP-2; JNK MAP kinase scaffold protein 2; JNK MAP kinase scaffold protein JIP2; JNK-interacting protein 2; Mitogen-activated protein kinase 8-interacting protein 2
Gene Aliases: IB-2; IB2; JIP2; MAPK8IP2; PRKM8IPL
UniProt ID: (Human) Q13387
Entrez Gene ID: (Human) 23542
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support