Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes an adhesion protein that plays a role in the organization of adherens junctions and tight junctions in epithelial and endothelial cells. The protein is a calcium(2+)-independent cell-cell adhesion molecule that belongs to the immunoglobulin superfamily and has 3 extracellular immunoglobulin-like loops, a single transmembrane domain, and a cytoplasmic region. This protein acts as a receptor for glycoprotein D of herpes simplex viruses 1 and 2, and pseudorabies virus and mediates viral entry into epithelial and neuronal cells. Mutations in this gene cause cleft lip and palate/ectodermal dysplasia 1 syndrome as well as non-syndromic cleft lip with or without cleft palate. Alternative splicing results in multiple transcript variants encoding proteins with distinct C-termini.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Jun NH2-terminal kinase-associated leucine zipper protein; C-Jun-amino-terminal kinase-interacting protein 4; Cancer/testis antigen 89; CT89; HLC-6; Human lung cancer oncogene 6 protein; JIP-4; JLP; JNK interacting protein; JNK-associated leucine-zipper protein; JNK-interacting leucine zipper; JNK-interacting protein 4; JNK/SAPK-associated protein; JNK/SAPK-associated protein 2; JSAP2; lung cancer oncogene 4; Max-binding protein; Mitogen-activated protein kinase 8-interacting protein 4; PHET; proliferation-inducing gene 6; Proliferation-inducing protein 6; Protein highly expressed in testis; sperm associated antigen 9 variant 5; Sperm surface protein; Sperm-associated antigen 9; Sperm-specific protein; Sunday driver 1
Gene Aliases: 3110018C07Rik; 4733401I23Rik; 4831406C20Rik; AW552012; CT89; HLC-6; HLC4; HLC6; HSS; JIP-4; JIP4; JLP; JSAP2; JSAP2a; KIAA0516; MAPK8IP4; PHET; PIG6; SPAG9; SYD1
UniProt ID: (Human) O60271, (Mouse) Q58A65
Entrez Gene ID: (Human) 9043, (Rat) 360600, (Mouse) 70834
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support