Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total JM4 protein.
JM4 is a 19 kDa four-transmembrane protein binding to the CCR5 receptor. It plays a role in the regulation of intracellular protein transport. It is expressed in normal brain, small intestine, lung, spleen, and pancreas and has also been detected in tumors of breast, colon, lung, and ovary.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Jena-Muenchen 4; PRA1 domain family 2; PRA1 domain family, member 2; PRA1 family protein 2
Gene Aliases: AA986329; C78013; DXImx39e; JM4; PRAF2; RGD1566287; Sfc20; Yip6a
UniProt ID: (Human) O60831, (Mouse) Q9JIG8
Entrez Gene ID: (Human) 11230, (Mouse) 54637, (Rat) 367743
Molecular Function: amino acid transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support