Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Active Motif
Histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a role in histone code. Does not demethylate histone H3 'Lys-4', H3 'Lys-27', H3 'Lys-36' nor H4 'Lys-20'. Only able to demethylate trimethylated H3 'Lys-9', with a weaker activity than KDM4A, KDM4C and KDM4D. Demethylation of Lys residue generates formaldehyde and succinate. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-trimethyl-L-lysine(9) demethylase 4B; JmjC domain-containing histone demethylation protein 3B; jumonji domain containing 2B; Jumonji domain-containing protein 2B; lysine (K)-specific demethylase 4B; Lysine-specific demethylase 4B; tudor domain containing 14B
Gene Aliases: JHDM3B; JMJD2B; KDM4B; KIAA0876; TDRD14B
UniProt ID: (Human) O94953
Entrez Gene ID: (Human) 23030
Molecular Function: histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support