Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
This antibody is predicted to react with Monkey, Pig, Cat, Sheep.
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Jumonji domain-containing protein 2C (JMJD2C)/ Lysine-specific demethylase 4C (KDM4C) is a histone demethylase that specifically demethylates 'Lys-9' and 'Lys-36' residues of histone H3; thereby, playing a central role in histone code. KDM4C does not demethylate histone H3 'Lys-4', H3 'Lys-27', or H4 'Lys-20'. It demethylates trimethylated H3 "Lys-9' and H3 "Lys-36' residues. There is no activity on mono- or dimethylated residues. Demethylation of Lys residues generates formaldehyde and succinate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-trimethyl-L-lysine(9) demethylase 4C; bA146B14.1; GASC-1 protein; Gene amplified in squamous cell carcinoma 1 protein; JmjC domain-containing histone demethylation protein 3C; Jumonji domain-containing protein 2C; lysine (K)-specific demethylase 4C; Lysine-specific demethylase 4C; RP11-169L18.1; tudor domain containing 14C
Gene Aliases: GASC1; JHDM3C; JMJD2C; KDM4C; KIAA0780; TDRD14C
UniProt ID: (Human) Q9H3R0
Entrez Gene ID: (Human) 23081
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support