Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Recommended positive controls: 293T, A375.
Predicted reactivity: Mouse (98%), Rat (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Covalent modification of histones plays critical role in regulating chromatin structure and transcription. While most covalent histone modifications are reversible, only recently has it been established that methyl groups are subject to enzymatic removal from histones. A family of novel JmjC domain-containing histone demethylation (JHDM) enzymes have been identified that perform this specific function. Histone demethylation by JHDM proteins requires cofactors Fe(II) and alpha-ketoglutarate. Family members include JHDM1 (demethylating histone 3 at lysine 36), and JHDM2A as well as JMJD2CH3K9 (both of which demethylate histone 3 at lysine 9). Contributions of histone demethylase activity to tumor development, decreases in cell proliferation, and hormone-dependent transcriptional activation have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-trimethyl-L-lysine(27) demethylase 6B; JmjC domain-containing protein 3; jumonji domain containing 3; jumonji domain containing 3, histone lysine demethylase; jumonji domain-containing 3; Jumonji domain-containing protein 3; lysine (K)-specific demethylase 6B; Lysine demethylase 6B; Lysine-specific demethylase 6B
Gene Aliases: 1700064E03Rik; BC038313; JMJD3; KDM6B; KIAA0346; mKIAA0346
UniProt ID: (Human) O15054, (Mouse) Q5NCY0
Entrez Gene ID: (Human) 23135, (Mouse) 216850, (Rat) 363630
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support