Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The antibody detects a 130 kDa* protein corresponding to the molecular mass of JMY on SDS-PAGE immunoblots of human HeLa, Jurkat cells, rat PC12 cells, and mouse brain and heart.
JMY acts both as a nuclear p53/TP53-cofactor and a cytoplasmic regulator of actin dynamics depending on conditions. In nucleus, acts as a cofactor that increases p53/TP53 response via its interaction with p300/EP300. Increases p53/TP53-dependent transcription and apoptosis, suggesting an important role in p53/TP53 stress response such as DNA damage. In cytoplasm, acts as a nucleation-promoting factor for both branched and unbranched actin filaments. Activates the Arp2/3 complex to induce branched actin filament networks. Also catalyzes actin polymerization in the absence of Arp2/3, creating unbranched filaments. Contributes to cell motility by controlling actin dynamics. May promote the rapid formation of a branched actin network by first nucleating new mother filaments and then activating Arp2/3 to branch off these filaments. The p53/TP53-cofactor and actin activator activities are regulated via its subcellular location.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ37870; Junction-mediating and -regulatory protein; junction-mediating and regulatory protein; p300 transcriptional cofactor JMY; similar to junction-mediating and regulatory protein; WAS protein homology region 2 domain containing 1-like 3
Gene Aliases: JMY; WHAMM2; WHDC1L3
UniProt ID: (Human) Q8N9B5, (Mouse) Q9QXM1
Entrez Gene ID: (Human) 133746, (Mouse) 57748, (Rat) 683593
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support