Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
JWA protein expression is affected by vitamin A and may be associated with the cytoskeleton. A protein similar to JWA in rats may play a role in the regulation of cell differentiation by inhibiting the cell membrane glutamate transporter EAAC1. The expression of the rat gene is upregulated by retinoic acid, which results in a specific reduction in EAAC1-mediated glutamate transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Addicsin; ADP-ribosylation factor-like protein 6-interacting protein 5; ADP-ribosylation-like factor 6 interacting protein 5; ARL-6-interacting protein 5; Glutamate transporter EAAC1-interacting protein; GTRAP3-18; PRA1 family protein 3; Prenylated Rab acceptor protein 2; Protein JWa
Gene Aliases: 5930404D22Rik; addiscin; Aip-5; Aip5; Arl6ip5; AV001879; Gtrap3-18; Jwa; Pra2; Praf3
UniProt ID: (Mouse) Q8R5J9
Entrez Gene ID: (Mouse) 65106
Molecular Function: amino acid transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support