Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Specificity: Specific to the NS1 protein. No cross reactivity with related viruses. Non-reactive with: TBE virus, Yellow Fever virus, West Nile virus, Dengue viruses (1-4), Zika virus and Chikungunya virus.
Best ELISA pairs (capture/conjugate): Japanese Encephalitis Virus NS1 Monoclonal antibody (Product # MA5-47513)/Japanese Encephalitis Virus NS1 Polyclonal antibody (Product # MA5-33269); Japanese Encephalitis Virus NS1 Monoclonal antibody (Product # MA5-47513)/Japanese Encephalitis Virus NS1 Monoclonal antibody (Product # MA5-47514); Japanese Encephalitis Virus NS1 Monoclonal antibody (Product # MA5-47513)/Japanese Encephalitis Virus NS1 Monoclonal antibody (Product # MA5-47515).
Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 µg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 ml. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Japanese encephalitis virus (JEV) is a flavivirus related to dengue, yellow fever and West Nile viruses, and is spread by mosquitoes. JEV is the main cause of viral encephalitis in many countries of Asia with an estimated 68000 clinical cases every year.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Japanese Encephalitis
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support