Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSLLMISENV KLAREYALLG NYDSAMVYYQ GVLDQMNKYL YSVKDTYLQQ KWQQVWQEIN VEAKHVKDIM KTLESFKLDS TPLKAAQHDL PASEGEVWSM PVPVERRPSP GPRKRQSSQY SDPKSHGNRP STTVRVHRSS AQNVHNDRGK AVRCREKKEQ NKGREEKGVL MVGPPGTGKT LLAKAVATEC KTTFFNVSSS TLTSKYRGES EKLVRLLFEM ARFYSPATIF IDEIDSICSR RGTSEEHEAS RRVKAELLVQ MDGVGGTSEN DDPSKMVMVL AATNFPWDID EALRRRLEKR IYIPLPSGMR P
Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. This gene encodes the p80 subunit. This protein is a member of the AAA family of ATPases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: katanin p60 (ATPase containing) subunit A 1; katanin p60 (ATPase-containing) subunit A 1; Katanin p60 ATPase-containing subunit A1; Katanin p60 subunit A1; KATNA1; p60 katanin
Gene Aliases: KATNA1
UniProt ID: (Human) O75449
Entrez Gene ID: (Human) 11104
Molecular Function: non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support