Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
KBTBD2 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB and kelch domain containing 1; BTB and kelch domain-containing protein 1; kelch repeat and BTB (POZ) domain containing 2; Kelch repeat and BTB domain-containing protein 2
Gene Aliases: BC022962; BKLHD1; CGI-73; KBTBD2; KIAA1489; mKIAA1489
UniProt ID: (Human) Q8IY47, (Mouse) G3X9X1
Entrez Gene ID: (Human) 25948, (Mouse) 210973, (Rat) 312372
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support