Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MSTQDERQIN TEYAVSLLEQ LKLFYEQQLF TDIVLIVEGT EFPCHKMVLA TCSSYFRAMF MSGLSESKQT HVHLRNVDAA TLQIIITYAY TGNLAMNDST VEQLYETACF LQVEDVLQRC REYLIKKINA ENCVRLLSFA DLFSCEELKQ SAKRMVEHKF TAVYHQDAFM QLSHDLLIDI LSSDNLNVGK EETVREAAML WLEYNTESRS QYLSSVLSQI RIDALSEVTQ RAWFQGLPPN DKSVVVQGLY KSMRKFFKPR LGMTKEEMMI FIEASSENPC SLYSSVCYSP QAEKVYKLCS RPADLHKVGT VVTPDNDIYI AGGQVPLKNT KTNHSKTSKL QTAFRTVNC
The KBTBD2 gene encodes the Kelch repeat and BTB (POZ) domain-containing protein 2, which is involved in various cellular processes, including protein ubiquitination and metabolic regulation. The KBTBD2 protein functions as a substrate adaptor for the Cullin-RING E3 ubiquitin ligase complex, facilitating the ubiquitination and subsequent proteasomal degradation of specific substrate proteins. This activity is crucial for maintaining cellular protein homeostasis and regulating metabolic pathways. KBTBD2 has been implicated in glucose metabolism and insulin signaling, where it plays a role in modulating the degradation of key regulatory proteins. Alterations in KBTBD2 function or expression are associated with metabolic disorders, such as obesity and type 2 diabetes. Additionally, KBTBD2's involvement in ubiquitin-mediated proteolysis underscores its significance in controlling various cellular responses and adaptations, making it a potential target for therapeutic interventions in metabolic diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB and kelch domain containing 1; BTB and kelch domain-containing protein 1; kelch repeat and BTB (POZ) domain containing 2; Kelch repeat and BTB domain-containing protein 2
Gene Aliases: BC022962; BKLHD1; CGI-73; KBTBD2; KIAA1489; mKIAA1489
UniProt ID: (Human) Q8IY47, (Mouse) G3X9X1
Entrez Gene ID: (Human) 25948, (Mouse) 210973, (Rat) 312372
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support