Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IMDAIKCVQG SGGLFPDARP STTEKYIFIH KTEENGENQY TFCYNIKSDS WKILPQSHLI DLPGSSLSSY GEKIFLTGGC KGKCCRTVRL HIAESYHDAT DQTWCY
Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
The BTB (Broad-Complex, Tram track and Bric a brac) domain, also known as the POZ (Poxvirus and Zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. KBTBD3 (kelch repeat and BTB domain-containing protein 3), also known as BKLHD3, is a 608 amino acid protein that contains one BACK (BTB/Kelch associated) domain, one BTB (POZ) domain and five kelch repeats. The gene encoding KBTBD3 maps to human chromosome 11, which houses over 1,400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB and kelch domain containing 3; BTB and kelch domain-containing protein 3; kelch repeat and BTB (POZ) domain containing 3; Kelch repeat and BTB domain-containing protein 3
Gene Aliases: BKLHD3; KBTBD3
UniProt ID: (Human) Q8NAB2
Entrez Gene ID: (Human) 143879
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support