Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: LESRVERHPL VRAQPELLRK VQMVKDAHEG RITTLRKKKK GKDGAGAKEA DKGTSKAKAE EDEEAERILP GILNDTLRFG MFL
Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.
The exact function of KBTBD5 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kelch repeat and BTB (POZ) domain containing 5; Kelch repeat and BTB domain-containing protein 5; kelch-like family member 40; Kelch-like protein 40; nemaline myopathy type 8; Sarcosynapsin
Gene Aliases: KBTBD5; KLHL40; NEM8; SRYP; SYRP
UniProt ID: (Human) Q2TBA0
Entrez Gene ID: (Human) 131377
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support