Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31781 has been tested in indirect ELISA.
KBTBD8 (Kelch Repeat And BTB Domain Containing 8) is a Protein Coding gene. Diseases associated with KBTBD8 include Treacher Collins Syndrome 1. Among its related pathways are Class I MHC mediated antigen processing and presentation and Innate Immune System. An important paralog of this gene is KBTBD2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kelch repeat and BTB (POZ) domain containing 8; Kelch repeat and BTB domain-containing protein 8; T-cell activation kelch repeat protein; TA-KRP
Gene Aliases: KBTBD8; KIAA1842; TA-KRP; TAKRP
UniProt ID: (Human) Q8NFY9
Entrez Gene ID: (Human) 84541
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support