Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide shares 91% with rat and mouse sequence.
Specificity of this antibody: KCNA3.
In neurons voltage-dependent potassium channels are key determinants of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGK5; HLK3; HPCN3; HUK3; KV3; MK3; potassium channel 3; potassium channel, voltage gated shaker related subfamily A, member 3; potassium voltage gated channel, shaker related subfamily, member 3; Potassium voltage-gated channel subfamily A member 3; potassium voltage-gated channel, shaker-related subfamily, member 3; RCK3; RGK5; type n potassium channel; Voltage-gated K(+) channel HuKIII; voltage-gated potassium channel protein Kv1.3; Voltage-gated potassium channel protein Kv1.3 (RGK5) (RCK3) (KV3); Voltage-gated potassium channel subunit Kv1.3; Voltage-gated potassium channel subunit Kv3
Gene Aliases: HGK5; HLK3; HPCN3; HUKIII; Kca1-3; KCNA3; KV1.3; Mk-3; MK3; PCN3
UniProt ID: (Human) P22001, (Mouse) P16390, (Rat) P15384
Entrez Gene ID: (Human) 3738, (Mouse) 16491, (Rat) 29731
Molecular Function:
voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support