Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MYPESTTGS PARLSLRQTG SPGMIYSTRY GSPKRQLQFY RNLGKSGLRV SCLGLGTWVT FGGQITDEMA EQLMTLAYDN GINLFDTAEV YAAGKAEVVL GNIIKKKGWR RSSLVITTKI FWGGKAETER GLSRKHIIEG LKASLERLQL EYVDVVFANR PDPNTPMEET VRAMTHVINQ GMAMYWGTSR WSSMEIMEAY SVARQFNLTP PICEQAEYHM FQREKVEVQL PELFHKIGVG AMTWSPLACG IVSGKYDSGI PPYSRASLKG YQWLKDKILS EEGRRQQAKL KELQAIAERL GCTLPQLAIA WCLRNEGVSS VLLGASNADQ LMENIGAIQV RVRGPAGQRA HPSPSPVQCI LPGSSCVPGS VLGTQDAPVN HQSCAPGELA FQQEQT (1-395 aa encoded by B C110351)
Kcnab2 is a voltage-gated potassium channels (KV) belong to the 6-TM family of potassium channel that also comprises the Ca2+-activated Slo (actually 7-TM) and the Ca2+-activated SK subfamilies. The alpha-subunits contain a single pore-forming region and combine to form tetramers. Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. The diverse functions of potassium channels include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). Kcnab2 is one of the beta subunits, which are auxiliary proteins associating with functional Kv-alpha subunits. Alternative splicing of the Kcnab2 gene results in multiple transcript variants encoding distinct isoforms. Diseases associated with KCNAB2 include Chromosome 1P36 Deletion Syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hKvbeta2; K(+) channel subunit beta-2; Kv-beta-2; Kvbeta2; Neuroimmune protein F5; potassium channel, voltage gated shaker-related subfamily A regulatory beta subunit 2; potassium channel, voltage gated subfamily A regulatory beta subunit 2; potassium voltage gated channel, shaker related subfamily, beta member 2; potassium voltage-gated channel, shaker-related subfamily, beta member 2; Voltage-gated potassium channel subunit beta-2
Gene Aliases: AKR6A5; Ckbeta2; F5; HKvbeta2; HKvbeta2.1; HKvbeta2.2; I2rf5; KCNA2B; KCNAB2; Kcnb3; KCNK2; KV-BETA-2
UniProt ID: (Human) Q13303, (Mouse) P62482, (Rat) P62483
Entrez Gene ID: (Human) 8514, (Mouse) 16498, (Rat) 29738
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support