Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MILSNTTAV TPFLTKLWQE TVQQGGNMSG LAHRSPRSGD GKLEALYVLM VLGFFGFFTL GIMLSYIRSK KLEHSNDPFN VYIESDAWQE KDKAYVQARV LESYRS (1-106 aa encoded by B C036452)
Kcne1 is a member of Voltage-gated potassium (Kv) channels that represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. The diverse functions of Kv channels include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The Kcne1 gene encodes a membrane protein which has sequence similarity to the KCNE1 gene product, a member of the potassium channel, voltage-gated, isk-related subfamily. Kcne1, an intronless gene is deleted in AMME contiguous gene syndrome and may be involved in the cardiac and neurologic abnormalities found in the AMME contiguous gene syndrome. Functionally, Kcne1 is a potassium channel ancillary subunit that is essential for generation of some native K(+) currents by virtue of formation of heteromeric ion channel complex with voltage-gated potassium (Kv) channel pore-forming alpha subunits. Kcne1 also functions as an inhibitory beta-subunit of the repolarizing cardiac potassium ion channel KCNQ1. Diseases associated with KCNE1 include Jervell And Lange-Nielsen Syndrome 2 and Long Qt Syndrome 5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiac delayed rectifier potassium channel protein; Delayed rectifier potassium channel subunit IsK; IKs producing slow voltage-gated potassium channel subunit beta Mink; JLNS 2; KCNE 1; KCNE1 (IsK); LQT 5; MGC33114; Minimal potassium channel; MinK; mISK; potassium (K+) channel protein, slowly activating (Isk); potassium channel, voltage gated subfamily E regulatory beta subunit 1; potassium voltage-gated channel Shaw-related subfamily member 1; Potassium voltage-gated channel subfamily E member 1; potassium voltage-gated channel, Isk-related family, member 1; potassium voltage-gated channel, Isk-related subfamily, member 1; voltage gated potassiun channel accessory subunit
Gene Aliases: ISK; JLNS; JLNS2; KCNE1; LQT2/5; LQT5; m; MinK; nmf190
UniProt ID: (Human) P15382, (Mouse) P23299, (Rat) P15383
Entrez Gene ID: (Human) 3753, (Mouse) 16509, (Rat) 25471
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support