Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Reconstitution: 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Human ether-a-go-go related gene (HERG) encodes the pore-forming alpha subunit of the delayed rectifier potassium channel IKr. There are two N-terminal splice variants of HERG include the full-length isoform 1 alpha and the shorter isoform 1 beta. Isoform 1 beta lacks the PAS motif and deactivates at a faster rate than isoform 1alpha. Residues within the C-terminal play a role in channel expression and channel gating, including voltage-dependent activation. HERG is expressed in the heart and is more abundantly expressed in the ventricles than in the atria. Mutations in the gene encoding HERG increase beat-to-beat variability and early after depolarization. These fluctuations facilitate the genesis and propagation of premature heartbeats associated with inheritable long QT syndrome type 2 and short QT syndrome type 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: Eag homolog; eag-related protein 1; ERG-1; Ether-a-go-go-related gene potassium channel 1; ether-a-go-go-related potassium channel protein; ether-a-go-go-related protein 1; HERG-1; M-erg1; potassium channel, voltage gated eag related subfamily H, member 2; Potassium voltage-gated channel subfamily H member 2; potassium voltage-gated channel, subfamily H (eag-related), member 2; r-ERG; RERG; Voltage-gated inwardly rectifying potassium channel KCNH2; Voltage-gated potassium channel subunit Kv11.1
Gene Aliases: ERG; ERG-1; ERG1; H-ERG; HERG; HERG1; KCNH2; Kv11.1; LQT2; SQT1
UniProt ID: (Human) Q12809, (Rat) O08962
Entrez Gene ID: (Human) 3757, (Rat) 117018
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support