Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 92%; Rat: 85%
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ELK channel 3; Ether-a-go-go-like potassium channel 3; Potassium voltage-gated channel subfamily H member 8; potassium voltage-gated channel, subfamily H, member 8; Voltage-gated potassium channel subunit Kv12.1
Gene Aliases: C130090D05Rik; ELK; ELK1; ELK3; Kcnh8; Kv12.1
UniProt ID: (Mouse) P59111
Entrez Gene ID: (Mouse) 211468
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support