Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This potassium channel is controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. This receptor plays a crucial role in regulating the heartbeat.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-activated inward rectifier potassium channel 1; GIRK-1; GIRK1; HGIRK1; Inward rectifier K(+) channel Kir3.1; inward rectifier K+ channel KIR3.1; KGA; KGB1; potassium channel inwarding rectifying channel subfamily J member 3; potassium channel subunit Kir3.1 type 3 delta; Potassium channel, inwardly rectifying subfamily J member 3; potassium channel, inwardly rectifying subfamily J, member 3; potassium inwardly-rectifying channel subfamily J member 3 splice variant 1e; potassium inwardly-rectifying channel, subfamily J, member 3; potassium voltage-gated channel subfamily J member 3
Gene Aliases: GIRK-1; GIRK1; Kcnf3; KCNJ3; KGA; KIR3.1
UniProt ID: (Human) P48549, (Mouse) P63250, (Rat) P63251
Entrez Gene ID: (Dog) 488355, (Human) 3760, (Mouse) 16519, (Rat) 50599
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support