Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: RLGLDQRAHS LDMLSPEKRS VFAALDTGRF KASSQESINN RPNNLRLKGP EQLNKHGQGA SEDNIINKFG STSRLTKRKN KDLKKTLPED VQKIYKTFRN YSLDEEKKEE ETEKMCNSDN SSTAMLTDCI QQHAELENGM IPTDTKDREP ENNSLLEDRN; Positive Samples: Mouse spleen, Rat testis; Cellular Location: Membrane, Multi-pass membrane protein
KCNK10 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The message for this gene is highly expressed in the kidney and pancreas. This channel is an open rectifier which primarily passes outward current under physiological K+ concentrations. The protein is stimulated strongly by arachidonic acid and to a lesser degree by membrane stretching, intracellular acidification, and general anaesthetics. Three transcript variants have been identified for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2P domain potassium channel TREK2; Outward rectifying potassium channel protein TREK-2; outward rectifying potassium channel TREK2; Potassium channel subfamily K member 10; potassium channel TREK-2; potassium channel, subfamily K, member 10; potassium channel, two pore domain subfamily K, member 10; protein phosphatase 1, regulatory subunit 97; TREK-2 K(+) channel subunit; TREK-2 two-pore-domain K+ channel; TWIK-related K+ channel 2
Gene Aliases: 1700024D23Rik; 3010005K24Rik; K2p10.1; KCNK10; PPP1R97; TREK-2; TREK2
UniProt ID: (Human) P57789, (Rat) Q9JIS4, (Mouse) Q8BUW1
Entrez Gene ID: (Human) 54207, (Rat) 65272, (Mouse) 72258
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support