Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in rat.
Specificity of this antibody: KCNK15.
This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel, however, it may require other non-pore-forming proteins for activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acid-sensitive potassium channel protein TASK-5; Potassium channel subfamily K member 15; potassium channel subfamily K member 15 (TASK-5); potassium channel, subfamily K, member 15 (TASK-5); rTASK-5; TWIK-related acid-sensitive K(+) channel 5
Gene Aliases: KCNK11; KCNK14; Kcnk15; KT3.3; TASK5
UniProt ID: (Rat) Q8R5I0
Entrez Gene ID: (Rat) 156873, (Mouse) 241769
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support