Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This antibody is 88% homologous to rat. It is not recommended for ICC/IF or Immunohistochemistry applications.
TREK-1 is a two-pore-domain background potassium channel expressed throughout the central nervous system. It is opened by polyunsaturated fatty acids and lysophospholipids. As well, it is regulated by various neurotransmitters. It has been shown that alterations in the functioning, regulation, or both of the TREK-1 channel may alter mood. TREK-1 is also activated by volatile anesthetics and has been suggested to be an important target in the action of these drugs. Therefore, this particular K+ channel emerges as a potential innovative target for developing new therapeutic agents for anesthesiology and neurology, such as antidepressants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: K2P2.1; K2P2.1 potassium channel; Kcnk2 potassium channel; Outward rectifying potassium channel protein TREK-1; Potassium channel subfamily K member 2; potassium channel subfamily k member 2 variant 1; potassium channel subfamily k member 2 variant 2; potassium channel, subfamily K, member 2; potassium channel, two pore domain subfamily K, member 2; potassium inwardly-rectifying channel, subfamily K, member 2; tandem-pore-domain potassium channel TREK-1; trek 1; TREK-1 K(+) channel subunit; TWIK-related potassium channel 1; two pore domain potassium channel TREK-1; Two pore domain potassium channel TREK1; Two pore potassium channel TPKC1; two-pore potassium channel 1
Gene Aliases: A430027H14Rik; AI848635; hTREK-1c; hTREK-1e; K2p2.1; KCNK2; TPKC1; TREK; TREK-1; TREK1
UniProt ID: (Human) O95069, (Mouse) P97438
Entrez Gene ID: (Bovine) 282590, (Human) 3776, (Mouse) 16526
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support