Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SRRTRAEMG GLTAQAASWT GTVTARVTQR AGPAAPPPEK EQPLLPPPPC PAQPLGRPRS PSPPEKAQPP SPPTASALDY PSENLAFIDE SSDTQSERGC PLPRAPRGRR RPNPPRKPVR PRGPGRPRDK GVPV (261-393 aa encoded by B C033577)
Potassium channels play a role in many cellular processes including maintenance of the action potential, muscle contraction, hormone secretion, osmotic regulation, and ion flow. This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The encoded protein homodimerizes and functions as an outwardly rectifying channel. It is expressed primarily in neural tissues and is stimulated by membrane stretch and polyunsaturated fatty acids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: K2P4.1 potassium channel; KT4.1b; Potassium channel subfamily K member 4; potassium channel, subfamily K, member 4; potassium channel, two pore domain subfamily K, member 4; potassium inwardly-rectifying channel subfamily K member 4; potassium inwardly-rectifying channel, subfamily K, member 4; protein expressed in male leptotene and zygotene spermatocytes 622; TRAAK; TWIK-related arachidonic acid-stimulated potassium channel protein; Two pore K(+) channel KT4.1; two pore K+ channel KT4.1; Two pore potassium channel KT4.1
Gene Aliases: K2p4.1; KCNK4; KT4.1; MLZ-622; Tex40; TRAAK; TRAAK1; TRAAKt
UniProt ID: (Human) Q9NYG8, (Mouse) O88454, (Rat) G3V8V5
Entrez Gene ID: (Human) 50801, (Mouse) 16528, (Rat) 116489
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support