Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Reconstitution: 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
KCNK6 channels play a critical role in maintaining the electrical stability of cells, and their dysfunction can contribute to a variety of pathophysiological conditions. KCNK6, also known as TWIK-2 (Tandem of Pore Domains in a Weak Inwardly Rectifying K+ Channel 2), is a member of the two-pore-domain potassium (K2P) channel family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ12282; Inward rectifying potassium channel protein TWIK-2; K2P6.1 potassium channel; Potassium channel subfamily K member 6; potassium channel, subfamily K, member 6; potassium channel, two pore domain subfamily K, member 6; potassium inwardly-rectifying channel, subfamily K, member 6; TWIK-originated similarity sequence; TWIK-originated sodium similarity sequence
Gene Aliases: K2p6.1; KCNK6; KCNK8; TOSS; TWIK-2; TWIK2
UniProt ID: (Human) Q9Y257, (Rat) G3V8R8
Entrez Gene ID: (Human) 9424, (Rat) 116491
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support