Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
Percent identity with other species by BLAST analysis: Human, Gorilla, Bovine (100%) Marmoset, Panda, Dog, Rabbit, Pig (93%) Mouse, Rat, Guinea pig (86%).
The protein encoded by this gene is part of a potentially heterotetrameric voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarization, which promotes calcium influx. The encoded protein may be part of the predominant calcium-activated potassium channel in T-lymphocytes. This gene is similar to other KCNN family potassium channel genes, but it differs enough to possibly be considered as part of a new subfamily.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gardos channel; HIK1; hKCa4; IKCa; IKCa1; intermediate conductance calcium-activated potassium channel protein 1; Intermediate conductance calcium-activated potassium channel protein 4; KCa3.1; KCNN 4; mIK1; mKCa4; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 4; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 4; putative erythrocyte intermediate conductance calcium-activated potassium Gardos channel; Putative Gardos channel; SK4; SKCa 4; SKCa4; small conductance calcium-activated potassium channel 4
Gene Aliases: DHS2; hIKCa1; hKCa4; hSK4; IK; IK1; IKCA1; KCa3.1; KCA4; KCNN4; mIKCa1; SK4; SKCas
UniProt ID: (Human) O15554, (Mouse) O89109
Entrez Gene ID: (Human) 3783, (Bovine) 534591, (Mouse) 16534
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support