Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MERKINRREK EKEYEGKHNS LEDTDQGKNC KSTLMTLNVG GYLYITQKQT LTKYPDTFLE GIVNGKILCP FDADGHYFID RDGLLFRHVL NFLRNGELLL PEGFRENQLL AQEAEFFQLK GLAEEVKSRW EKEQLTPRET TFLEITDNHD RSQGLRIFCN APDFISKIKS RIVLVSKSRL DGFPEEFSIS SNIIRFKYFI KSENGTRLVL KEDNTFVCTL ETLKFEAIMM ALKCGFRLLT SLDCSKGSIV HSDALHFIK
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB/POZ domain-containing protein KCTD4; potassium channel tetramerisation domain containing 4
Gene Aliases: bA321C24.3; KCTD4
UniProt ID: (Human) Q8WVF5
Entrez Gene ID: (Human) 386618
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support