Product References
Dependence of ABCB1 transporter expression and function on distinct sphingolipids generated by ceramide synthases-2 and -6 in chemoresistant renal cancer.
The Journal of biological chemistry
Lee WK,Maaß M,Quach A,Poscic N,Prangley H,Pallott EC,Kim JL,Pierce JS,Ogretmen B,Futerman AH,Thévenod F
PA1-013 was used in Immunocytochemistry to conclude that a critical balance in ceramide/SM species is prerequisite to ABCB1 expression and functionalization, which could be targeted to reverse multidrug resistance in renal cancers.
Tue Feb 01 00:00:00 UTC 2022
Induction of Axonal Outgrowth in Mouse Hippocampal Neurons via Bacterial Magnetosomes.
International journal of molecular sciences
De Vincentiis S,Falconieri A,Mickoleit F,Cappello V,Schüler D,Raffa V
PA1-013 was used in Immunocytochemistry-immunoflourescence to data show that bacterial magnetosomes are more efficient than synthetic iron oxide nanoparticles in stimulating axonal outgrowth via SG.
Fri Apr 16 00:00:00 UTC 2021
Induction of Axonal Outgrowth in Mouse Hippocampal Neurons via Bacterial Magnetosomes.
International journal of molecular sciences
De Vincentiis S,Falconieri A,Mickoleit F,Cappello V,Schüler D,Raffa V
PA1-013 was used in Immunocytochemistry-immunoflourescence to data show that bacterial magnetosomes are more efficient than synthetic iron oxide nanoparticles in stimulating axonal outgrowth via SG.
Fri Apr 16 00:00:00 UTC 2021
Prohibitin 1 is essential to preserve mitochondria and myelin integrity in Schwann cells.
Nature communications
Della-Flora Nunes G,Wilson ER,Marziali LN,Hurley E,Silvestri N,He B,O'Malley BW,Beirowski B,Poitelon Y,Wrabetz L,Feltri ML
PA1-013 was used in Immunohistochemistry to elucidate the functional contribution of prohibitin 1 in schwann cells.
Wed Jun 02 00:00:00 UTC 2021
Prohibitin 1 is essential to preserve mitochondria and myelin integrity in Schwann cells.
Nature communications
Della-Flora Nunes G,Wilson ER,Marziali LN,Hurley E,Silvestri N,He B,O'Malley BW,Beirowski B,Poitelon Y,Wrabetz L,Feltri ML
PA1-013 was used in Immunohistochemistry to elucidate the functional contribution of prohibitin 1 in schwann cells.
Wed Jun 02 00:00:00 UTC 2021
Posttranslational modifications of serine protease TMPRSS13 regulate zymogen activation, proteolytic activity, and cell surface localization.
The Journal of biological chemistry
Martin CE,Murray AS,Sala-Hamrick KE,Mackinder JR,Harrison EC,Lundgren JG,Varela FA,List K
Published figure using KDEL polyclonal antibody (Product # PA1-013) in Immunocytochemistry
Fri Oct 01 00:00:00 UTC 2021
Dopamine Transporter Localization in Medial Forebrain Bundle Axons Indicates Its Long-Range Transport Primarily by Membrane Diffusion with a Limited Contribution of Vesicular Traffic on Retromer-Positive Compartments.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Bagalkot TR,Block ER,Bucchin K,Balcita-Pedicino JJ,Calderon M,Sesack SR,Sorkin A
PA1-013 was used in Immunohistochemistry to suggest that DAT traffics between midbrain and striatum mainly by lateral diffusion in the plasma membrane with only a limited contribution of vesicular transport in recycling endosomes.
Wed Jan 13 00:00:00 UTC 2021
Analysis of Intracellular Vesicles in B Lymphocytes: Antigen Traffic in the Spotlight.
Methods in molecular biology (Clifton, N.J.)
Hernández-Pérez S,Runsala M,Šuštar V,Mattila PK
PA1-013 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot to provide a protocol for 3D analysis of the intracellular vesicle traffic, in either fixed or living cells, using B lymphocytes (B cells) as a model system.
Wed Aug 11 00:00:00 UTC 2021
Ankyrin Is An Intracellular Tether for TMC Mechanotransduction Channels.
Neuron
Tang YQ,Lee SA,Rahman M,Vanapalli SA,Lu H,Schafer WR
PA1-013 was used in Immunocytochemistry to identify UNC-44/ankyrin as an essential component of the TMC-1 mechanotransduction channel complex in the sensory cilia of Caenorhabditis elegans mechanoreceptor neurons.
Wed Jul 08 00:00:00 UTC 2020
The Perinuclear ER Scales Nuclear Size Independently of Cell Size in Early Embryos.
Developmental cell
Mukherjee RN,Sallé J,Dmitrieff S,Nelson KM,Oakey J,Minc N,Levy DL
PA1-013 was used in Immunocytochemistry to monitor nuclear dynamics in early sea urchin embryos and find that nuclei undergo substantial growth in each interphase, reaching a maximal size prior to mitosis that declined steadily over the course of development.
Mon Aug 10 00:00:00 UTC 2020
Extremely Low Forces Induce Extreme Axon Growth.
The Journal of neuroscience : the official journal of the Society for Neuroscience
De Vincentiis S,Falconieri A,Mainardi M,Cappello V,Scribano V,Bizzarri R,Storti B,Dente L,Costa M,Raffa V
PA1-013 was used in Immunocytochemistry to generate a dragging force using a protocol based on magnetic nanoparticles (NPs).
Wed Jun 24 00:00:00 UTC 2020
Extremely Low Forces Induce Extreme Axon Growth.
The Journal of neuroscience : the official journal of the Society for Neuroscience
De Vincentiis S,Falconieri A,Mainardi M,Cappello V,Scribano V,Bizzarri R,Storti B,Dente L,Costa M,Raffa V
PA1-013 was used in Immunocytochemistry to generate a dragging force using a protocol based on magnetic nanoparticles (NPs).
Wed Jun 24 00:00:00 UTC 2020
TUBB1 mutations cause thyroid dysgenesis associated with abnormal platelet physiology.
EMBO molecular medicine
Stoupa A,Adam F,Kariyawasam D,Strassel C,Gawade S,Szinnai G,Kauskot A,Lasne D,Janke C,Natarajan K,Schmitt A,Bole-Feysot C,Nitschke P,Léger J,Jabot-Hanin F,Tores F,Michel A,Munnich A,Besmond C,Scharfmann R,Lanza F,Borgel D,Polak M,Carré A
Published figure using KDEL polyclonal antibody (Product # PA1-013) in Immunohistochemistry
Sat Dec 01 00:00:00 UTC 2018
TUBB1 mutations cause thyroid dysgenesis associated with abnormal platelet physiology.
EMBO molecular medicine
Stoupa A,Adam F,Kariyawasam D,Strassel C,Gawade S,Szinnai G,Kauskot A,Lasne D,Janke C,Natarajan K,Schmitt A,Bole-Feysot C,Nitschke P,Léger J,Jabot-Hanin F,Tores F,Michel A,Munnich A,Besmond C,Scharfmann R,Lanza F,Borgel D,Polak M,Carré A
Published figure using KDEL polyclonal antibody (Product # PA1-013) in Immunohistochemistry
Sat Dec 01 00:00:00 UTC 2018
CIB2 interacts with TMC1 and TMC2 and is essential for mechanotransduction in auditory hair cells.
Nature communications
Giese APJ,Tang YQ,Sinha GP,Bowl MR,Goldring AC,Parker A,Freeman MJ,Brown SDM,Riazuddin S,Fettiplace R,Schafer WR,Frolenkov GI,Ahmed ZM
PA1-013 was used in Immunocytochemistry-immunoflourescence to study the exact functions of calcium and integrin-binding protein 2.
Thu Jun 29 00:00:00 UTC 2017
Axons provide the secretory machinery for trafficking of voltage-gated sodium channels in peripheral nerve.
Proceedings of the National Academy of Sciences of the United States of America
González C,Cánovas J,Fresno J,Couve E,Court FA,Couve A
PA1-013 was used in immunocytochemistry to characterize the secretory machinery in axons and study its contribution to plasma membrane delivery of sodium channels
Tue Feb 16 00:00:00 UTC 2016
Hyaluronan Rafts on Airway Epithelial Cells.
The Journal of biological chemistry
Abbadi A,Lauer M,Swaidani S,Wang A,Hascall V
Published figure using KDEL polyclonal antibody (Product # PA1-013) in Immunocytochemistry
Fri Jan 15 00:00:00 UTC 2016
Hyaluronan Rafts on Airway Epithelial Cells.
The Journal of biological chemistry
Abbadi A,Lauer M,Swaidani S,Wang A,Hascall V
Published figure using KDEL polyclonal antibody (Product # PA1-013) in Immunocytochemistry
Fri Jan 15 00:00:00 UTC 2016
Primary murine airway smooth muscle cells exposed to poly(I,C) or tunicamycin synthesize a leukocyte-adhesive hyaluronan matrix.
The Journal of biological chemistry
Lauer ME,Mukhopadhyay D,Fulop C,de la Motte CA,Majors AK,Hascall VC
PA1-013 was used in western blot to study the response of primary murine airway smooth muscle cells to poly (I,C) or tunicamycin
Fri Feb 20 00:00:00 UTC 2009
Primary murine airway smooth muscle cells exposed to poly(I,C) or tunicamycin synthesize a leukocyte-adhesive hyaluronan matrix.
The Journal of biological chemistry
Lauer ME,Mukhopadhyay D,Fulop C,de la Motte CA,Majors AK,Hascall VC
PA1-013 was used in western blot to study the response of primary murine airway smooth muscle cells to poly (I,C) or tunicamycin
Fri Feb 20 00:00:00 UTC 2009
Differentiated murine airway epithelial cells synthesize a leukocyte-adhesive hyaluronan matrix in response to endoplasmic reticulum stress.
The Journal of biological chemistry
Lauer ME,Erzurum SC,Mukhopadhyay D,Vasanji A,Drazba J,Wang A,Fulop C,Hascall VC
Published figure using KDEL polyclonal antibody (Product # PA1-013) in Immunocytochemistry
Fri Sep 19 00:00:00 UTC 2008
Differentiated murine airway epithelial cells synthesize a leukocyte-adhesive hyaluronan matrix in response to endoplasmic reticulum stress.
The Journal of biological chemistry
Lauer ME,Erzurum SC,Mukhopadhyay D,Vasanji A,Drazba J,Wang A,Fulop C,Hascall VC
Published figure using KDEL polyclonal antibody (Product # PA1-013) in Immunocytochemistry
Fri Sep 19 00:00:00 UTC 2008
Differentiated murine airway epithelial cells synthesize a leukocyte-adhesive hyaluronan matrix in response to endoplasmic reticulum stress.
The Journal of biological chemistry
Lauer ME,Erzurum SC,Mukhopadhyay D,Vasanji A,Drazba J,Wang A,Fulop C,Hascall VC
Published figure using KDEL polyclonal antibody (Product # PA1-013) in Immunocytochemistry
Fri Sep 19 00:00:00 UTC 2008
Differentiated murine airway epithelial cells synthesize a leukocyte-adhesive hyaluronan matrix in response to endoplasmic reticulum stress.
The Journal of biological chemistry
Lauer ME,Erzurum SC,Mukhopadhyay D,Vasanji A,Drazba J,Wang A,Fulop C,Hascall VC
Published figure using KDEL polyclonal antibody (Product # PA1-013) in Immunocytochemistry
Fri Sep 19 00:00:00 UTC 2008
Further characterization of mammalian ceramide kinase: substrate delivery and (stereo)specificity, tissue distribution, and subcellular localization studies.
Journal of lipid research
Van Overloop H,Gijsbers S,Van Veldhoven PP
PA1-013 was used in immunocytochemistry to investigate related factors affecting recombinant human ceramide kinase's activity
Wed Feb 01 00:00:00 UTC 2006
Changes of endoplasmic reticulum chaperone complexes, redox state, and impaired protein disulfide reductase activity in misfolding alpha1-antitrypsin transgenic mice.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Papp E,Száraz P,Korcsmáros T,Csermely P
PA1-013 was used in western blot to investigate the changes of endoplasmic reticulum chaperone complexes, redox state, and impaired protein disulfide reductase activity in misfolding alpha1-antitrypsin transgenic mice
Mon May 01 00:00:00 UTC 2006
Functional proteomic screens reveal an essential extracellular role for hsp90 alpha in cancer cell invasiveness.
Nature cell biology
Eustace BK,Sakurai T,Stewart JK,Yimlamai D,Unger C,Zehetmeier C,Lain B,Torella C,Henning SW,Beste G,Scroggins BT,Neckers L,Ilag LL,Jay DG
PA1-013 was used in immunoprecipitation to study the role of extracellular Hsp90-alpha in the activation of MMP2 and invasiveness of fibrosarcoma cells
Tue Jun 01 00:00:00 UTC 2004
Protein-disulfide isomerase-mediated reduction of the A subunit of cholera toxin in a human intestinal cell line.
The Journal of biological chemistry
Orlandi PA
PA1-013 was used in western blot to study the mechanisms of cholera toxin A subunit reduction in a human intestinal cell line
Fri Feb 14 00:00:00 UTC 1997