Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. 자세히 알아보기
FIGURE: 1 / 1
A 1:1,000 dilution of MA5-45876 was sufficient for detection of KDEL in 10 µg of HeLa cell lysates by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Detects KDEL tagged proteins.
The sequence Lys-Asp-Glu-Leu (KDEL) or a closely related sequence, is present at the carboxy-terminus of soluble endoplasmic reticulum (ER) resident proteins and some membrane proteins. 78 and 94 kDa glucose regulated proteins, GRP78 and GRP94 respectively, and protein disulfide isomerase (PDI) all share the C-terminal KDEL sequence. The presence of carboxy-terminal KDEL appears to be necessary for ER retention and appears to be sufficient to reduce the secretion of proteins from the ER. This retention is reported to be mediated by a KDEL receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 78 kDa glucose-regulated protein; 94 kDa glucose-regulated protein; 94 kDa glucose-regulated protein {ECO:0000250|UniProtKB:P08113}; Binding-immunoglobulin protein; BiP; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; Endoplasmic reticulum resident protein 99; Endoplasmin; endoplasmin {ECO:0000250|UniProtKB:P08113}; endothelial cell (HBMEC) glycoprotein; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; epididymis secretory sperm binding protein Li 89n; ER lumen protein retaining receptor 1; ER lumen protein-retaining receptor 1; ERp99; glucose regulated protein, 78 kDa; glucose-regulated protein, 78kDa; gp96 homolog; GRP 78; GRP-78; GRP-94; heat shock 70 kDa protein 5; heat shock 70kD protein 5; heat shock 70kD protein 5 (glucose-regulated protein, 78kD); heat shock 70kDa protein 5 (glucose-regulated protein); heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa); Heat shock protein 70 family protein 5; Heat shock protein 90 kDa beta member 1; heat shock protein 90 kDa beta member 1 {ECO:0000250|UniProtKB:P08113}; heat shock protein 90kDa beta (Grp94), member 1; heat shock protein 90kDa beta family member 1; Heat shock protein family A member 5; Heat shock protein family C member 4; HSP70 family protein 5; hsp90b1 {ECO:0000250|UniProtKB:P08113}; Immunoglobulin heavy chain-binding protein; KDEL; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1; KDEL endoplasmic reticulum protein retention receptor 1; KDEL receptor 1; Lys-Asp-Glu-Leu; Polymorphic tumor rejection antigen 1; Putative MAPK-activating protein PM23; RP23-446N16.1; Steroidogenesis-activator polypeptide; stress-inducible tumor rejection antigen gp96; transforming growth factor alpha regulated gene 2; tumor rejection antigen (gp96) 1; Tumor rejection antigen 1; Tumor rejection antigen gp96; XAP-1 antigen
Gene Aliases: 8030486F04Rik; AL022860; AU019543; AW215843; baffled; BIP; D2Wsu141e; D2Wsu17e; ECGP; endoplasmin; ERD2; ERD2.1; ERp99; GP96; GRP78; GRP94; HDEL; HEL-S-125m; HEL-S-89n; HEL35; Hsce70; HSP90B1; HSPA5; HSPC4; KDELR1; mBiP; MIF2; PM23; SEZ-7; Sez7; TA-3; Targ2; Tra-1; TRA1
UniProt ID: (Human) P14625, (Human) P11021, (Mouse) P08113, (Rat) Q66HD0, (Rat) P06761, (Mouse) P20029, (Human) P24390, (Mouse) Q99JH8, (Rat) Q569A6
Entrez Gene ID: (Human) 7184, (Human) 3309, (Mouse) 22027, (Rat) 362862, (Rat) 25617, (Mouse) 14828, (Human) 10945, (Mouse) 68137, (Rat) 361577
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support