Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Store as a concentrated solution. Centrifuge briefly prior to opening vial.
Retention of resident soluble proteins in the lumen of the endoplasmic reticulum (ER) is achieved in both yeast and animal cells by their continual retrieval from the cis-Golgi, or a pre-Golgi compartment. Sorting of these proteins is dependent on a C-terminal tetrapeptide signal, usually lys-asp-glu-leu (KDEL) in animal cells, and his-asp-glu-leu (HDEL) in S. cerevisiae. This process is mediated by a receptor that recognizes, and binds the tetrapeptide-containing protein, and returns it to the ER. In yeast, the sorting receptor encoded by a single gene, ERD2, which is a seven-transmembrane protein. Unlike yeast, several human homologs of the ERD2 gene, constituting the KDEL receptor gene family, have been described. The protein encoded by this gene was the first member of the family to be identified, and it encodes a protein structurally and functionally similar to the yeast ERD2 gene product.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2; CG5183-PA; CG5183-PB; CG5183-PC; ELP-1; ER lumen protein retaining receptor; ER lumen protein retaining receptor 1; ER lumen protein retaining receptor 2; ER lumen protein retaining receptor 3; ER lumen protein-retaining receptor; ER lumen protein-retaining receptor 1; ER lumen protein-retaining receptor 2; ER lumen protein-retaining receptor 3; ERD-2-like protein; ERD2-like protein 1; ERD23; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 2; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 3; KDEL endoplasmic reticulum protein retention receptor 1; KDEL endoplasmic reticulum protein retention receptor 2; KDEL endoplasmic reticulum protein retention receptor 3; KDEL receptor 1; KDEL receptor 2; KDEL receptor 3; KDEL receptor-like protein; KDEL-receptor; KdelR-PA; KdelR-PB; KdelR-PC; lethal (2) 31Em; Putative MAPK-activating protein PM23
Gene Aliases: 1110007A14Rik; 8030486F04Rik; AI173274; AW215843; BEST:CK00230; CG5183; CK00230; Dmel\CG5183; Dmel_CG5183; dmErd2; ELP-1; ERD2; ERD2.1; ERD2.2; ERD23; ERD2L3; ergr15; HDEL; I79_001653; I79_020931; I79_023963; KDELR; KDELR1; KDELR2; KDELR3; l(2)31Em; l(2)k00311; PM23; RCJMB04_8l23; XELAEV_18045328mg
UniProt ID: (Bovine) P33946, (Human) P24390, (Bovine) Q2KJ37, (Human) P33947, (Chicken) Q5ZKX9, (Human) O43731, (Fruit fly) O76767, (Mouse) Q99JH8, (Rat) Q569A6, (Mouse) Q9CQM2, (Rat) Q5U305, (Mouse) Q8R1L4, (Xenopus) Q7ZXS5
Entrez Gene ID: (Pig) 100520962, (Bovine) 618184, (Sheep) 101122047, (Human) 10945, (Dog) 484406, (Rabbit) 100349892, (Pig) 100625446, (Bovine) 531676, (Human) 11014, (Chicken) 771927, (Dog) 612839, (Sheep) 101116624, (Rabbit) 100338871, (Chicken) 418026, (Rabbit) 100338388, (Dog) 474509, (Pig) 100514523, (Sheep) 101121418, (Bovine) 509687, (Human) 11015, (Fruit fly) 34427, (Chinese hamster) 100750912, (Mouse) 68137, (Rat) 361577, (Mouse) 66913, (Rat) 304290, (Chinese hamster) 100765889, (Mouse) 105785, (Chinese hamster) 100750636, (Rat) 315131, (Xenopus) 380330
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support