Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
1 µg/mL was sufficient for detection of KDEL receptor in 20 µg monkey Vero cell lysate by colorimetric immunoblot analysis using Goat Anti-Mouse IgG:AP as the secondary.|Detects approximately 25kDa.
KDELR1 (KDEL Endoplasmic Reticulum Protein Retention Receptor 1) is a Protein Coding gene. Diseases associated with KDELR1 include Syndromic X-Linked Intellectual Disability 94 and Myopathy, Centronuclear, 1. Among its related pathways are Transport to the Golgi and subsequent modification and Activation of cAMP-Dependent PKA. Gene Ontology (GO) annotations related to this gene include ER retention sequence binding and KDEL sequence binding. An important paralog of this gene is KDELR2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ER lumen protein retaining receptor 1; ER lumen protein-retaining receptor 1; KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1; KDEL endoplasmic reticulum protein retention receptor 1; KDEL receptor 1; KDEL receptor-like protein; Putative MAPK-activating protein PM23
Gene Aliases: 8030486F04Rik; AW215843; ERD2; ERD2.1; HDEL; I79_020931; KDELR; KDELR1; PM23
UniProt ID: (Human) P24390, (Bovine) P33946, (Mouse) Q99JH8, (Rat) Q569A6
Entrez Gene ID: (Human) 10945, (Pig) 100520962, (Bovine) 618184, (Rabbit) 100349892, (Sheep) 101122047, (Dog) 484406, (Mouse) 68137, (Rat) 361577, (Chinese hamster) 100750912
Molecular Function: membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support