Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MDLDRASLDL MIRLLELEQD ASSAKLLNEK DMNKIKEKIR RLCETVHNKH LDLENITTGH LAMETLLSLT SKRAGDWFKE ELRLLGGLDH IVDKVKECVD HLSRDEDEEK LVASLWGAER CLRVLESVTV HNPENQSYLI AYKDSQLIVS SAKALQHCEE LIQQYNRAED SICLADSKPL PHQNVTNHVG KAVEDCMRAI IGVLLNLTND NEWGSTKTGE QDGLIGTALN CVLQVPKYLP QEQRFDIRVL LFLERERAAQ LAESKTDELI KDAPTTQHDK SGEWQETSGE IQWVSTEKTD GTEEKHKKEE EDEELDLNKA LQHAGKHMED CIVASYTALL LGCLCQESPI NVTTVREYLP EGDFSIMTEM LKKFLSFMNL TCAVGTTGQK SISRVIEYLE HC
Transcriptional repressor which may play a role in development of the central nervous system (CNS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: friend of EBNA2 (Epstein-Barr virus nuclear protein 2); Friend of EBNA2 protein; WAPL cohesin release factor; Wings apart-like protein homolog
Gene Aliases: FOE; KIAA0261; WAPAL; WAPL
UniProt ID: (Human) Q7Z5K2
Entrez Gene ID: (Human) 23063
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support