Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MFEEPEWAEA APVAAGLGPV ISRPPPAASS QNKGSKRRQL LATLRALEAA SLSQHPPSLC ISDSEEEEEE RKKKCPKKAS FASASAEVGK KGKKKCQKQG PPCSDSEEEV ERKKKCHKQA LVGSDSAEDE KRKRKCQKHA PINSAQHLDN VDQTGPKAWK GSTTNDPPKQ SPGSTSPKPP HTLSRKQWRN RQKNKRRCKN KFQPPQVPDQ APAEAPTEKT EVSPVPRTDS HEARAGALRA RMAQRLDGAR FRYLNEQLYS GPSSAAQRLF QEDPEAFLLY HRGFQSQVKK WPLQPVDRIA RDLRQRPASL VVADFGCGDC RLASSIRNPV HCFDLASLDP RVTVCDMAQV PLEDESVDVA VFCLSLMGTN IRDFLEEANR VLKPGGLLKV AEVSSRFEDV RTFLRAVTKL GFKIVSKDLT NSHFFLFDFQ KTGPPLVGPK AQLSGLQLQP CLYKRR
Essential component of the eNoSC (energy-dependent nucleolar silencing) complex, a complex that mediates silencing of rDNA in response to intracellular energy status and acts by recruiting histone-modifying enzymes. The eNoSC complex is able to sense the energy status of cell: upon glucose starvation, elevation of NAD(+)/NADP(+) ratio activates SIRT1, leading to histone H3 deacetylation followed by dimethylation of H3 at 'Lys-9' (H3K9me2) by SUV39H1 and the formation of silent chromatin in the rDNA locus. In the complex, RRP8 binds to H3K9me2 and probably acts as a methyltransferase. Its substrates are however unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cerebral protein 1; HUCE1; hucep1; Nucleomethylin; Ribosomal RNA-processing protein 8; RRP-8; RRP8 methyltransferase homolog
Gene Aliases: hucep-1; KIAA0409; NML; RRP8
UniProt ID: (Human) O43159
Entrez Gene ID: (Human) 23378
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support