Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLTTLKPFGS VSVESKMNNK AGSFFWNLRQ FSTLVSTSRT MRLCCLGLCK PKIVHSNWNI LNNFHNRMQS TDIIRYLFQD AFIFKSDVGF QTKGISTLTA
This gene encodes a protein that is localized in the mitochondrial inner compartment and that may play a role in mitochondrial apoptosis. Nonsense mutations have been reported to result in cytochrome c oxidase deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FAST kinase domain-containing protein 2; FAST kinase domain-containing protein 2, mitochondrial
Gene Aliases: FASTKD2; KIAA0971
UniProt ID: (Human) Q9NYY8
Entrez Gene ID: (Human) 22868
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support