Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQLDFKSIHV QEPQEPSLQD ASLKPKVECS VVTEFTDHIC VTMDAELIMF LHDLVSAYLK EKEKAIFPPR ILSTRPGQKS PIIIHDDNSS DKDREDSITY TTVDWRDFMC NTWHLEPTLR LISWTGRKID PVGVDYILQK LGFHHARTTI PKWLQRGVMD PLDKVLSVLI KKLGTALQDE KEKKGKDKEE H
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bridge-like lipid transfer protein family member 1; Fragile site-associated protein
Gene Aliases: BLTP1; FSA; KIAA1109; KIAA1371; TWEEK
UniProt ID: (Human) Q2LD37
Entrez Gene ID: (Human) 84162
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support