Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MASRQPEVPA LEASAPLGKM SLPIGIYRRA VSYDDTLEDP APMTPPPSDM GSVPWKPVIP ERKYQHLAKV EEGEASLPSP AMTLSSAIDS VDKVPVVKAK ATHVIMNSLI TKQTQESIQH FERQAGLRDA GYTPHKGLTT EETKYLRVAE ALHKLKLQSG EVTKEERQPA SAQSTPSTTP HSSPKQRPRG WFTSGSSTAL PGPNPSTMDS GSGDKDRNLS DKWSLFGPRS LQKYDSGSFA TQAYRGAQKP SPLELIRAQA NRMAEDPAAL KPPKMDIPVM EGKKQPPRAH NLKPRDLNVL TPTGF
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-derived rescue factor p60MONOX; Flavin monooxygenase motif-containing protein of 33 kDa; flavine monooxygenase motif-containing protein of 33 kDa; Putative monooxygenase p33MONOX
Gene Aliases: KIAA1191; P33MONOX; p60MONOX
UniProt ID: (Human) Q96A73
Entrez Gene ID: (Human) 57179
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support