Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LSEERSCREE LGQQHPRHSR PRKTAASPEK PQTTKATGRM NSHLAPPEKR KGRPEPSTKS GGGRCAIHPR RSKGADLERS NPLVAAVGEI GLV
Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 34%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CEP295 N-terminal-like protein; differential display clone 8; KIAA1731 N-terminal like protein
Gene Aliases: CEP295NL; DDC8; KIAA1731NL
UniProt ID: (Human) Q96MC4
Entrez Gene ID: (Human) 100653515
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support