Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAALLRRLLQ RERPSAASGR PVGRREANLG TDAGVAVRVR FAPSPTGFLH LGGLRTALYN YIFAKKYQGS FILRLEDTDQ TRVVPGAAEN IEDMLEWAGI PPDESPRRGG PAGPYQQSQR LELYAQATEA LLKTGAAYPC FCSPQRLELL KKEALRNHQT PRYDNRCRNM SQEQVAQKLA KDPKPAIRFR LEQVVPAFQD LVYGWNRHEV ASVEGDPVIM KSDGFPTYHL ACVVDDHHMG ISHVLRGSEW LVSTAKHLLL YQALGWQPPH FAHLPLLLNR DGSKLSKRQG DVFLEHFAAD GFLPDSLLDI ITNCGSGFAE NQMGRTLPEL ITQFNLTQVT CHSALLDLEK LPEFNRLHLQ RLVSNESQRR QLVGKLQVLV EEAFGCQLQN RDVLNPVYVE RILLLRQGHI CRLQDLVSPV YSYLWTRPAV GRAQLDAISE KVDVIAKRVL GLLERSGMSL TQDMLNGELK KLSEGLEGTK YSNVMKLLRM ALSGQQQGPP VAEMMLALGP KEVRERIQKV VSS
Glutamyl-tRNA synthetase (GluRS or EARS2) a class I aminoacyl-tRNA synthetase (aaRS), is primarily responsible for the glutamylation of tRNAGlu. It is part of the "minimal set" of seventeen aaRSs found in every living organism and its presence is essential for the viability of cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GluRS; glutamate tRNA ligase 2, mitochondrial; glutamate--tRNA ligase; Glutamate--tRNA(Gln) ligase EARS2, mitochondrial; Glutamyl-tRNA synthetase; Mitochondrial glutamyl-tRNA synthetase; mtGluRS; Nondiscriminating glutamyl-tRNA synthetase EARS2, mitochondrial; probable glutamyl-tRNA synthetase, mitochondrial
Gene Aliases: COXPD12; EARS2; gluRS; KIAA1970; MSE1
UniProt ID: (Human) Q5JPH6
Entrez Gene ID: (Human) 124454
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support