Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SDRAIQSLT IQTACAFEQL VVLMKHWLSD LLPCTNIARL EDELRQEVKK LGGYLQLFLQ GCCLDISSMI KEAQKNAIQI VQQAVKYVGQ LAVLKGSKLH FLENGNNKAA SVQEEFMDAV CDGVGLGMKI LLDSGLEKAK ELQHELFRQC TKNEVTKEMK TNAMGLIRSL ENIFAESKIK SFRRQVQEEN FEYQDFKRMV NRAPEFLKLK HCLEKAIEII ISALKGCHSD INLLQTCVES IRNLASDFYS DFSVPSTSVG SYESRVTHIV HQELESLAKS LLFCFESEES PDLLKPWETY NQNTKEEHQQ SKSSGIDGSK NKGVPKRVYE LHGSSAAVSS EECTPSRIQW V (1299-1648 aa encoded by BC113742)
This gene encodes a member of the kinesin-3 superfamily of microtubule motor proteins. These proteins are involved in numerous processes including vesicle transport, chromosome segregation, mitotic spindle formation, and cytokinesis. In human HeLa-S3 and 293T cells, this protein is localized to the cytoplasm during interphase, to the spindle poles and spindle microtubules during mitosis, and to the midbody during cytokinesis. An internal motor domain displays microtubule-dependent ATPase activity, consistent with its function as a microtubule motor protein. Knockdown of this gene results in failed cytokinesis with endoreplication, which results in multinucleated cells. This gene has been identified as a likely oncogene in breast, lung and ovarian cancers, as well as retinoblastomas and gliomas. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kinesin-like protein KIF14
Gene Aliases: KIAA0042; KIF14; MKS12
UniProt ID: (Human) Q15058
Entrez Gene ID: (Human) 9928
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support