Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSLHSTHNRN NSGDILDIPS SQNSSSLNAL THSSRLKLHL KSDMSECEND DPLLRSAGKV RDINRTYVIS ASRKTADMPL TPNPVGRLAL QRRTTRNKES SLLVSELEDT TEKTAETRLT LQRRAKTDSA EKWKTAEIDS VKMTLNVGGE TENNGVSKES RTNVRIVNNA KNSFVASSVP LDEDPQVIEM MADKKYKETF SAPSRANENV ALKYSSNRPP IASLSQTEVV RSGHLTTKPT QSKLDIKVLG TGNLYHRSIG KEIAKTSNKF GSLEKRTPTK CTTEHKLTTK CSLPQLKSPA PSILKNRMSN LQVKQRPKSS FLANKQERSA ENTILPEEET VVQNTSAGKD PLKVENSQVT VAVRVRPFTK REKIEKASQV VFMSGKEITV EHPDTKQVYN FIYDVSFWSF DECHPHYASQ TTVYEKLAAP LLERAFEGFN TCLFAYGQTG SGKSYTMMGF SEEPGIIPRF CEDLFSQVAR KQTQEVSYHI EMSFFEVYNE KIHDLLVCKD ENGQRKQPLR VREHPVYGPY VEALSMNIVS SYADIQSWLE LGNKQRATAA TGMNDKSSRS HSVFTLVMTQ TKTEFVEGEE HDHRITSRIN LIDLAGSERC STAHTNGDRL KEGVSINKSL LTLGKVISAL SEQANQRSVF IPYRESVLTW LLKESLGGNS KTAMIATISP AASNIEETLS TLRYANQARL IVNIAKVNED MNAKLIRELK AEIAKLKAAQ RNSRNIDPER YRLCRQEITS LRMKLHQQER DMAEMQRVWK EKFEQAEKRK LQETKELQKA GIMFQMDNHL PNLVNLNEDP QLSEMLLYMI KEGTTTVGKY KPNSSHDIQL SGVLIADDHC TIKNFGGTVS IIPVGEAKTY VNGKHILEIT VLRHGDRVIL GGDHYFRFNH PVEVQKGKRP SGRDTPISEG PKDFEFAKNE LLMAQRSQLE AEIKEAQLKA KEEMMQGIQI AKEMAQQELS SQKAAYESKI KALEAELREE SQRKKMQEIN NQKANHKIEE LEKAKQHLEQ EIYVNKKRLE METLATKQAL EDHSIRHARI LEALETEKQK IAKEVQILQQ NRNNRDKTFT VQTTWSSMKL SMMIQEANAI SSKLKTYYVF GRHDISDKSS SDTSIRVRNL KLGISTFWSL EKFESKLAAM KELYESNGSN RGEDAFCDPE DEWEPDITDA PVSSLSRRRS RSLMKNRRIS GCLHDIQVHP IKNLHSSHSS GLMDKSSTIY SNSAESFLPG ICKELIGSSL DFFGQSYDEE RTIADSLINS FLKIYNGLFA ISKAHEEQDE ESQDNLFSSD RAIQSLTIQT ACAFEQLVVL MKHWLSDLLP CTNIARLEDE LRQEVKKLGG YLQLFLQGCC LDISSMIKEA QKNAIQIVQQ AVKYVGQLAV LKGSKLHFLE NGNNKAASVQ EEFMDAVCDG VGLGMKILLD SGLEKAKELQ HELFRQCTKN EVTKEMKTNA MGLIRSLENI FAESKIKSFR RQVQEENFEY QDFKRMVNRA PEFLKLKHCL EKAIEIIISA LKGCHSDINL LQTCVESIRN LASDFYSDFS VPSTSVGSYE SRVTHIVHQE LESLAKSLLF CFESEESPDL LKPWETYNQN TKEEHQQSKS SGIDGSKNKG VPKRVYELHG SSPAVSSEEC TPSRIQWV
This gene encodes a member of the kinesin-3 superfamily of microtubule motor proteins. These proteins are involved in numerous processes including vesicle transport, chromosome segregation, mitotic spindle formation, and cytokinesis. In human HeLa-S3 and 293T cells, this protein is localized to the cytoplasm during interphase, to the spindle poles and spindle microtubules during mitosis, and to the midbody during cytokinesis. An internal motor domain displays microtubule-dependent ATPase activity, consistent with its function as a microtubule motor protein. Knockdown of this gene results in failed cytokinesis with endoreplication, which results in multinucleated cells. This gene has been identified as a likely oncogene in breast, lung and ovarian cancers, as well as retinoblastomas and gliomas. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kinesin-like protein KIF14
Gene Aliases: KIAA0042; KIF14; MKS12
UniProt ID: (Human) Q15058
Entrez Gene ID: (Human) 9928
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support