Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: LENGNNKAAS VQEEFMDAVC DGVGLGMKIL LDSGLEKAKE LQHELFRQCT KNEVTKEMKT NAMGLIRSLE NIFAESKIKS FRRQVQEENF EYQDFKRMVN RAPEFLKLKH CLEKAIEIII SALKGCHSDI NLLQTCVESI RNLASDFYSD FSVPSTSVGS YESRVTHIVH QELESLAKSL LFCFESEESP DLLKPWETYN QNTKEEHQQS KSSGIDGSKN KGVPKRVYEL HGSSPAVSSE ECTPSRIQWV; Positive Samples: HeLa, HL-60; Cellular Location: Cytoplasm, Midbody, Nucleus, cytoskeleton, spindle
This gene encodes a member of the kinesin-3 superfamily of microtubule motor proteins. These proteins are involved in numerous processes including vesicle transport, chromosome segregation, mitotic spindle formation, and cytokinesis. In human HeLa-S3 and 293T cells, this protein is localized to the cytoplasm during interphase, to the spindle poles and spindle microtubules during mitosis, and to the midbody during cytokinesis. An internal motor domain displays microtubule-dependent ATPase activity, consistent with its function as a microtubule motor protein. Knockdown of this gene results in failed cytokinesis with endoreplication, which results in multinucleated cells. This gene has been identified as a likely oncogene in breast, lung and ovarian cancers, as well as retinoblastomas and gliomas. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kinesin-like protein KIF14; N-3 kinesin; truncated kinesin family member 14
Gene Aliases: D1Ertd367e; E130203M01; KIAA0042; KIF14; MKS12
UniProt ID: (Human) Q15058, (Mouse) L0N7N1
Entrez Gene ID: (Human) 9928, (Mouse) 381293, (Rat) 360849
Molecular Function: microtubule binding motor protein non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support