Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MASEAVKVV VRCRPMNQRE RELRCQPVVT VDCARAQCCI QNPGAADEPP KQFTFDGAYH VDHVTEQIYN EIAYPLVEGV TEGYNGTIFA YGQTGSGKSF TMQGLPDPPS QRGIIPRAFE HVFESVQCAE NTKFLVRASY LEIYNEDVRD LLGADTKQKL ELKEHPEKGV YVKGLSMHTV HSVAQCEHIM ETGWKNRSVG YTLMNKDSSR SHSIFTISIE MSAVDERGKD HLRAGKLNLV DLAGSERQSK TGATGERLKE ATKINLSLSA LGNVISALVD GRCKHVPYRD SKLTRLLQDS LGGNTKTLMV ACLSPADNNY DETLSTLRYA NRAKNIRNKP RINEDPKDAL LR (1-351 aa encoded by BC065927)
Dendrite-specific motor protein which, in association with the Apba1-containing complex (LIN-10-LIN-2-LIN-7 complex), transports vesicles containing N-methyl-D-aspartate (NMDA) receptor subunit NR2B along microtubules. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIF17 variant protein; KIF3-related motor protein; kinesin 17; kinesin family member 17; Kinesin-like protein KIF17; MmKIF17; N-4 kinesin
Gene Aliases: 5930435E01Rik; AW492270; KIAA1405; KIF17; KIF17B; KIF3X; KLP-2; mKIAA1405; OSM-3; RGD1562511
UniProt ID: (Human) Q9P2E2, (Mouse) Q99PW8
Entrez Gene ID: (Human) 57576, (Rat) 500571, (Mouse) 16559
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support