Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
May have a role in a signaling pathway. Could act as a tumor suppressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kinesin-like protein KIF18A; Marrow stromal KIF18A; MS-KIF18A; protein phosphatase 1, regulatory subunit 99
Gene Aliases: KIF18A; MS-KIF18A; OK/SW-cl.108; PPP1R99
UniProt ID: (Human) Q8NI77
Entrez Gene ID: (Human) 81930
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support